Etylklorformiat 99%, AcroSeal®
Leverantör: Thermo Scientific
Fara
Synonyms: Chloroformic acid ethyl ester
430791000 430798000
430791000.EA 745 SEK
430791000. 430798000.
Etylklorformiat 99%, AcroSeal®
Etylklorformiat
Formel: C₃H₅ClO₂
MW: 108,52 g/mol
Kokpunkt: 94 °C (760 mmHg)
Smältpunkt: –81 °C
Densitet: 1,135 g/cm³
Flampunkt: 10 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000644
CAS nummer: 541-41-3
UN: 1182
ADR: 6.1,I
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3940 to 1.3960 (20°C, 589 nm)
GC ≤1.0 % Diethyl carbonate
Color scale ≤50 APHA
Free acid (HCl) ≤0.1 %