1-Cyclohexenylboronic acid pinacol ester 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-Cyclohexeneboronic acid pinacol ester, 2-(Cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Cyclohexene-1-boronic acid pinacol ester
430310010 430315000
ACRO430310010EA 4400 SEK
ACRO430310010 ACRO430315000
1-Cyclohexenylboronic acid pinacol ester 97%
1-Cyclohexenylboronic acid pinacol ester
Formel: C₁₂H₂₁BO₂
MW: 208.1049 g/mol
Kokpunkt: 125 °C (.2 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD05663845
CAS nummer: 141091-37-4
Order Now
Specifikation testresultat
Appearance Clear colorless to tan Liquid
Infrared spectrum Conforms
GC ≥95 %