1H-Indazole-6-carboxylic acid 97%
Leverantör: Thermo Scientific
Varning
430120010 430120050
ACRO430120010EA 1225 SEK
ACRO430120010 ACRO430120050
1H-Indazole-6-carboxylic acid 97%
1H-Indazole-6-carboxylic acid
Formel: C₈H₆N₂O₂
MW: 162,1454 g/mol
Smältpunkt: 297…300 °C
Storage Temperature: Omgivande
MDL Number: MFCD06804571
CAS nummer: 704-91-6
Order Now
Specifikation testresultat
Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with HClO4 ≥95.0 %