Tetrabutylammonium hydroxide 1 M i metanol, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
428951000 428958000
428951000.EA 1400 SEK
428951000. 428958000.
Tetrabutylammonium hydroxide 1 M i metanol, AcroSeal®
Tetrabutylammonium hydroxide
Formel: C₁₆H₃₇NO
MW: 259.48 g/mol
Kokpunkt: 65 °C (760 mmHg)
Smältpunkt: –98 °C
Densitet: 0.83 g/cm³
Flampunkt: 12 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009425
CAS nummer: 2052-49-5
UN: 3286
ADR: 3 II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Solution
Titration with HCl 0.95 to 1.10 M