ω-Bromo-3-hydroxyacetophenone 96%
Leverantör: Thermo Scientific
Fara
Synonyms: α-Bromo-3-hydroxyacetophenone, 3-Hydroxyphenacyl bromide, 2-Bromo-1-(3-hydroxyphenyl)ethan-1-one, 2-Bromo-1-(3-hydroxyphenyl)ethanone, 2-Bromo-3'-hydroxyacetophenone
428620010 428620050
ACRO428620010EA 1675 SEK
ACRO428620010 ACRO428620050
ω-Bromo-3-hydroxyacetophenone 96%
ω-Bromo-3-hydroxyacetophenone
Formel: C₈H₇BrO₂
MW: 215,05 g/mol
Smältpunkt: 70…74 °C
Storage Temperature: Omgivande
MDL Number: MFCD05664326
CAS nummer: 2491-37-4
UN: 3261
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Brown Powder
Infrared spectrum Conforms
HPLC ≥95 %
NMR (1H-NMR) Consistent with structure
Specific optical rotation -22° to -26° (20°C, 589 nm) (c=1, ethanol)