Cyclopropylboronic acid pinacol ester 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-Cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
428590010 428590050
428590010.EA 915 SEK
428590010. 428590050.
Cyclopropylboronic acid pinacol ester 95%
Cyclopropylboronic acid pinacol ester
Formel: C₉H₁₇BO₂
MW: 168.04 g/mol
Kokpunkt: 146 °C (760 mmHg)
Densitet: 0.922 g/cm³
Flampunkt: 40 °C
Storage Temperature: Kylskåp
MDL Number: MFCD05663847
CAS nummer: 126689-01-8
UN: 3272
ADR: 3 III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
NMR (1H-NMR) Consistent with structure