EGTA-Na4 (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt) 95%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecane-1,14-dioic acid, tetrasodium salt, Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt, EGTA tetrasodium
428570100 428570500
ACRO428570100EA 1700 SEK
ACRO428570100 ACRO428570500
EGTA-Na4 (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt) 95%
EGTA-Na4 (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt)
Formel: C₁₄H₂₀N₂Na₄O₁₀
MW: 468,28 g/mol
Smältpunkt: 54…56 °C
Storage Temperature: Omgivande
MDL Number: MFCD00274420
CAS nummer: 13368-13-3
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Assay 18.46 to 20.82 % (ICP on dried sample)
Loss on drying ≤20 % (110°C, 16 hrs)