Methyl cis-11-eicosenoate 99%
Leverantör: Thermo Scientific
Varning
Synonyms: cis-11-Eicosenoic acid methyl ester, methyl (Z)-icos-11-enoate, (Z)-Methyl icos-11-enoate, (11Z)-11-Icosenoic acid methyl ester
428530010 428530050
ACRO428530010EA 5500 SEK
ACRO428530010 ACRO428530050
Methyl cis-11-eicosenoate 99%
Methyl cis-11-eicosenoate
Formel: C₂₁H₄₀O₂
MW: 324,549 g/mol
Smältpunkt: –15 °C
Flampunkt: >113 °C
Storage Temperature: Frys
MDL Number: MFCD00010458
CAS nummer: 2390-09-2
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure