Tris(trimethylsilyl)phosphite 92%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Tris(trimethylsilyl)phosphite
428520050 428520250 428521000
ACRO428520050EA 1300 SEK
ACRO428520050 ACRO428520250 ACRO428521000
Tris(trimethylsilyl)phosphite 92%
Tris(trimethylsilyl)phosphite
Formel: C₉H₂₇O₃PSi₃
MW: 298.54 g/mol
Kokpunkt: 78…81 °C (8.3 mmHg)
Densitet: 0.893 g/cm³
Flampunkt: 66 °C
Storage Temperature: Omgivande
MDL Number: MFCD00015588
CAS nummer: 1795-31-9
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4080 to 1.4110 (20°C, 589 nm)
GC ≥90 %