2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2,8,9-Tris(2-methylpropyl)-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane, Triisobutylphosphatrane, 2,8,9-Tri-i-butyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane, [Nitrilotrisethylenetris(isobutylimino)]phosphine
428250010 428250050
ACRO428250010EA 1625 SEK
ACRO428250010 ACRO428250050
2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane 97%
2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane
Formel: C₁₈H₃₉N₄P
MW: 342.51 g/mol
Kokpunkt: 110…118 °C (.5 mmHg)
Densitet: 0.964 g/cm³
Flampunkt: 68 °C
Storage Temperature: Omgivande
MDL Number: MFCD03840347
CAS nummer: 331465-71-5
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Elemental Analysis 15.70 to 17.01 % N