N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14) 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-(N,N-Dimethylmyristylammonio)propanesulphonate, Tetradecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner Salt
427740050 427740250
ACRO427740050EA 335 SEK
ACRO427740050 ACRO427740250
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14) 97%
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14)
Formel: C₁₉H₄₁NO₃S
MW: 363,5988 g/mol
Smältpunkt: 246…248 °C
Storage Temperature: Omgivande
MDL Number: MFCD00036910
CAS nummer: 14933-09-6
Order Now
Specifikation testresultat
Appearance White Powder
Infrared spectrum Conforms
Elemental Analysis 3.72 to 3.99 % N