2-metyl-2-buten 90% balance 2-Methyl-1-butene
Leverantör: Thermo Scientific
Fara
Synonyms: Trimethylethylene, Isoamylene, Isopentene
414090010 414090025 414092500
ACRO414090010EA 6075 SEK
ACRO414090010 ACRO414090025 ACRO414092500
2-metyl-2-buten 90% balance 2-Methyl-1-butene
2-metyl-2-buten
Formel: C₅H₁₀
MW: 70,13 g/mol
Kokpunkt: 35…38 °C (760 mmHg)
Smältpunkt: –134 °C
Densitet: 0,66 g/cm³
Flampunkt: –45 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009276
CAS nummer: 513-35-9
UN: 2460
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥88 % (Balance mainly 2-methyl-1-butene)
Titration with NaOH 96.0 to 104.0 %