5-Aminofluorescein (isomer I), ren
Leverantör: Thermo Scientific
Varning
Synonyms: 5-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, Fluoresceinamine, isomer I, 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one
400770010 400770050
ACRO400770010EA 1925 SEK
ACRO400770010 ACRO400770050
5-Aminofluorescein (isomer I), ren
5-Aminofluorescein (isomer I)
Formel: C₂₀H₁₃NO₅
MW: 347,33 g/mol
Smältpunkt: 223 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005052
CAS nummer: 3326-34-9
Order Now
Specifikation testresultat
Appearance Dark red-brown Crystalline powder
Infrared spectrum Conforms
HPLC 95 % Typical
GC ≥97.5 %