((3-Bromopropoxy)methyl)benzene 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Benzyl 3-bromopropyl ether, 3-bromopropylbenzyl ether
399130010 399130100
ACRO399130010EA 760 SEK
ACRO399130010 ACRO399130100
((3-Bromopropoxy)methyl)benzene 98%
((3-Bromopropoxy)methyl)benzene
Formel: C₁₀H₁₃BrO
MW: 229,117 g/mol
Kokpunkt: 130…132 °C (8 mmHg)
Densitet: 1,298 g/cm³
Flampunkt: 113 °C
Storage Temperature: Omgivande
MDL Number: MFCD00134570
CAS nummer: 54314-84-0
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5300 to 1.5330 (20°C, 589 nm)
GC ≥97.5 %