4-Fluoro-2-methoxyphenol 97%
Leverantör: Thermo Scientific
Varning
398990010 398990050
ACRO398990010EA 495 SEK
ACRO398990010 ACRO398990050
4-Fluoro-2-methoxyphenol 97%
4-Fluoro-2-methoxyphenol
Formel: C₇H₇FO₂
MW: 142,13 g/mol
Kokpunkt: 195 °C (760 mmHg)
Densitet: 1,247 g/cm³
Flampunkt: 101 °C
Storage Temperature: Omgivande
MDL Number: MFCD00070797
CAS nummer: 450-93-1
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5180 (20°C, 589 nm)
GC ≥96.0 %