1-Butanol, extra dry 99+%, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol
398960010 398961000
398960010.EA 2950 SEK
398960010. 398961000.
1-Butanol, extra dry 99+%, AcroSeal®
1-Butanol
Formel: H₃C(CH₂)₂CH₂OH
Kokpunkt: 117,6 °C (760 mmHg)
Smältpunkt: –89 °C
Densitet: 0,81 g/cm³
Flampunkt: 35 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002964
CAS nummer: 71-36-3
UN: 1120
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.001 %