O-tert-Butylhydroxylamine hydrochloride 99%
Leverantör: Thermo Scientific
Varning
Synonyms: O-(tert-Butyl)hydroxylamine hydrochloride
399680010 399680050
ACRO399680010EA 2000 SEK
ACRO399680010 ACRO399680050
O-tert-Butylhydroxylamine hydrochloride 99%
O-tert-Butylhydroxylamine hydrochloride
Formel: C₄H₁₂ClNO
MW: 125,5972 g/mol
Smältpunkt: 155 °C
Storage Temperature: Omgivande
MDL Number: MFCD00043272
CAS nummer: 39684-28-1
UN: 1325
ADR: 4.1,III
Order Now
Specifikation testresultat
Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Titration Argentometric 98.5 to 101.5 %