5-(Trifluoromethoxy)salicylaldehyde 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Hydroxy-5-(trifluoromethoxy)benzaldehyde
398200010 398202500
ACRO398200010EA 3600 SEK
ACRO398200010 ACRO398202500
5-(Trifluoromethoxy)salicylaldehyde 99%
5-(Trifluoromethoxy)salicylaldehyde
Formel: C₈H₅F₃O₃
MW: 206,12 g/mol
Kokpunkt: 82 °C (60 mmHg)
Smältpunkt: 31…33 °C
Storage Temperature: Omgivande
MDL Number: MFCD00075249
CAS nummer: 93249-62-8
Order Now
Specifikation testresultat
Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 30 to 34 °C
HPLC ≥98.5 %
NMR (1H-NMR) Consistent with structure