Bis(2-metoxietyl)eter, extra dry 99+%, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
397210010 397211000
397210010.EA 6625 SEK
397210010. 397211000.
Bis(2-metoxietyl)eter, extra dry 99+%, AcroSeal®
Bis(2-metoxietyl)eter
Formel: C₆H₁₄O₃
Kokpunkt: 162 °C (760 mmHg)
Smältpunkt: –64 °C
Densitet: 0,937 g/cm³
Flampunkt: 51 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008503
CAS nummer: 111-96-6
UN: 3271
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.001 %