Klorbenzen, extra dry 99.8%, AcroSeal®
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
Phenylchloride, Benzene Chloride, Monochlorobenzene
396970010
396971000
396970010.EA
3875
SEK
396970010.
396971000.
Klorbenzen, extra dry 99.8%, AcroSeal®
Klorbenzen
Formel:
C₆H₅Cl MW: 112,56 g/mol Kokpunkt: 131 °C (760 mmHg) Smältpunkt: –45 °C Densitet: 1,1075 g/cm³ Flampunkt: 23 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00000530 CAS nummer: 108-90-7 UN: 1134 ADR: 3,III |
Specifikation testresultat
Appearance | Clear colorless Liquid |
Water | ≤0.005 % (Coulometric) |
GC | ≥99.8 % |
Residue after evaporation | ≤0.001 % |
Filtered to 0.2 micrometer | Filtered |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...