Benzylalkohol, extra dry 98+%, AcroSeal®
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol
396880010 396881000
396880010.EA 6175 SEK
396880010. 396881000.
Benzylalkohol, extra dry 98+%, AcroSeal®
Benzylalkohol
Formel: C₆H₅CH₂OH
MW: 108.14 g/mol
Kokpunkt: 205 °C (760 mmHg)
Smältpunkt: –15 °C
Densitet: 1.044 g/cm³
Flampunkt: 96 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004599
CAS nummer: 100-51-6
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥98.0 %
Residue after evaporation ≤0.05 %