4-Bromo-1H-pyrazole 98%
Leverantör: Thermo Fisher Scientific
Varning
396820010 396820050 396820250
ACRO396820010EA 805 SEK
ACRO396820010 ACRO396820050 ACRO396820250
4-Bromo-1H-pyrazole 98%
4-Bromo-1H-pyrazole
Formel: C₃H₃BrN₂
Kokpunkt: 250…260 °C (760 mmHg)
Smältpunkt: 93…96 °C
Storage Temperature: Omgivande
MDL Number: MFCD00075602
CAS nummer: 2075-45-8
Order Now
Specifikation testresultat
Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 92 to 99 °C
HPLC ≥97.5 %