1-Dodekansulfonsyra natriumsalt, vattenfri 99+% för jonparkromatografi
Leverantör: Thermo Fisher Scientific
Synonyms: Sodium 1-dodecanesulphonate, Dodecane-1-sulphonic acid sodium salt, Sodium 1-dodecansulphonate, 1-Dodecanesulphonic acid, sodium salt, Sodium dodecane-1-sulphonate
396720050 396720250
ACRO396720050EA 3175 SEK
ACRO396720050 ACRO396720250
1-Dodekansulfonsyra natriumsalt, vattenfri 99+% för jonparkromatografi
1-Dodekansulfonsyra natriumsalt
Formel: C₁₂H₂₅NaO₃S
MW: 272.38 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007527
CAS nummer: 2386-53-0
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 %
UV at 230 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.18 M soln.) Passes test
Suitability (HPLC gradient) Passes test