Natriumpentan-1-sulfonatmonohydrat, vattenfri 99+% för jonparkromatografi
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate
396650250 396651000 396655000
ACRO396650250EA 8550 SEK
ACRO396650250 ACRO396651000 ACRO396655000
Natriumpentan-1-sulfonatmonohydrat, vattenfri 99+% för jonparkromatografi
Natriumpentan-1-sulfonatmonohydrat
Formel: C₅H₁₁O₃SNa
MW: 174.2 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007541
CAS nummer: 22767-49-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb1c23c8a6fe3c156a3aacb62721e1d1a34efb0&vid=1936dcdb47ba374ce21fd758660f7ca6ba95d254&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 % (150°C)
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test