Sodium tetrachloroaurate hydrate 98%
Leverantör: Thermo Fisher Scientific
Varning
396270010 396270050
ACRO396270010EA 3200 SEK
ACRO396270010 ACRO396270050
Sodium tetrachloroaurate hydrate 98%
Sodium tetrachloroaurate hydrate
Formel: AuCl₄Na·H₂O
Storage Temperature: Omgivande
MDL Number: MFCD00149161
CAS nummer: 29156-65-8
UN: 1759
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Yellow to orange to golden Powder or crystals or granules
Assay 48.3 to 50.3 % (Au)