4-Methoxy-2-methylphenylboronic acid 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-Methoxy-2-methylbenzeneboronic acid, (4-Methoxy-2-methylphenyl)boronic acid
393850010 393850050
ACRO393850010EA 1075 SEK
ACRO393850010 ACRO393850050
4-Methoxy-2-methylphenylboronic acid 97%
4-Methoxy-2-methylphenylboronic acid
Formel: C₈H₁₁BO₃
Smältpunkt: 148…161 °C
Storage Temperature: Omgivande
MDL Number: MFCD02684315
CAS nummer: 208399-66-0
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
HPLC ≥96.0 %