2-(Trifluoromethoxy)phenylboronic acid 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-Trifluoromethoxyphenylboronic acid, 2-(Trifluoromethoxy)benzeneboronic acid, (2-(Trifluoromethoxy)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethoxy)phenyl)borane
393810010 393810050
ACRO393810010EA 900 SEK
ACRO393810010 ACRO393810050
2-(Trifluoromethoxy)phenylboronic acid 97%
2-(Trifluoromethoxy)phenylboronic acid
Formel: C₇H₆BF₃O₃
MW: 205.93 g/mol
Smältpunkt: 121…125 °C
Storage Temperature: Omgivande
MDL Number: MFCD01320763
CAS nummer: 175676-65-0
Order Now
Specifikation testresultat
Appearance White to light brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %