Kalium molybdat (VI) 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Potassium molybdate, Molybdic acid potassium salt, Dipotassium tetraoxomolybdate, Molybdic acid, potassium salt, Potassium molybdenum oxide, di-Potassium molybdate
393231000 393235000
ACRO393231000EA 1275 SEK
ACRO393231000 ACRO393235000
Kalium molybdat (VI) 95%
Kalium molybdat (VI)
Formel: K₂MoO₄
MW: 238.13 g/mol
Smältpunkt: 919 °C
Densitet: 2.91
Storage Temperature: Omgivande
MDL Number: MFCD00011370
CAS nummer: 13446-49-6
Merck Index: 13,07732
Order Now
Specifikation testresultat
Appearance White Powder and/or chunks
Assay ≥94 %
Chloride (Cl) ≤0.05 %
Phosphate (PO4) ≤0.03 %
Sulfate (SO4) ≤0.05 %
Iron (Fe) ≤0.01 %