5-Methylsalicylaldehyde 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Hydroxy-5-methylbenzaldehyde
392630010 392630050
ACRO392630010EA 695 SEK
ACRO392630010 ACRO392630050
5-Methylsalicylaldehyde 99%
5-Methylsalicylaldehyde
Formel: C₈H₈O₂
MW: 136,15 g/mol
Kokpunkt: 217 °C (760 mmHg)
Smältpunkt: 54…57 °C
Densitet: 1,0913 g/cm³ (59 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00274297
CAS nummer: 613-84-3
Order Now
Specifikation testresultat
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 57 °C
GC ≥98.5 %