4-Cyanophenylhydrazine hydrochloride 97+%
Leverantör: Thermo Fisher Scientific
Varning
392300010 392300050 392300250
ACRO392300010EA 445 SEK
ACRO392300010 ACRO392300050 ACRO392300250
4-Cyanophenylhydrazine hydrochloride 97+%
4-Cyanophenylhydrazine hydrochloride
Formel: C₇H₈ClN₃
Smältpunkt: 249 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00673994
CAS nummer: 2863-98-1
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Light orange to brown Powder
Infrared spectrum Conforms
HPLC ≥97.0 %