Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Leverantör: Thermo Scientific
Varning
392140010 392140050
ACRO392140010EA 2275 SEK
ACRO392140010 ACRO392140050
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Formel: C₅₅H₄₅ClOP₃Ru
MW: 951,405 g/mol
Smältpunkt: 204…206 °C
Storage Temperature: Omgivande
MDL Number: MFCD00049804
CAS nummer: 16971-33-8
Order Now
Specifikation testresultat
Appearance Cream to tan to brown Powder or crystals
Infrared spectrum Conforms
Ruthenium (Ru) ≥10.3 %