(3-Chloropropyl)triethoxysilane 97+%
Leverantör: Thermo Fisher Scientific
Varning
391890010 391890050 391890250
ACRO391890010EA 2625 SEK
ACRO391890010 ACRO391890050 ACRO391890250
(3-Chloropropyl)triethoxysilane 97+%
(3-Chloropropyl)triethoxysilane
Formel: ClH₂CCH₂CH₂Si(OCH₂CH₃)₃
Kokpunkt: 230 °C (760 mmHg)
Densitet: 1,004 g/cm³
Flampunkt: 78 °C
Storage Temperature: Omgivande
MDL Number: MFCD00018985
CAS nummer: 5089-70-3
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4210 (20°C, 589 nm)
GC ≥97.0 %