Natriumpolyfosfat (Grahams salt), ren
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Metaphosphoric acid, sodium salt, Sodium hexametaphosphate, Sodium polymetaphosphate, Polyphosphate sodium salt, Graham′s salt, Sodium metaphosphate, SHMP
390930010 390930050 390932500
ACRO390930010EA 1025 SEK
ACRO390930010 ACRO390930050 ACRO390932500
Natriumpolyfosfat (Grahams salt), ren
Natriumpolyfosfat (Grahams salt)
Formel: (NaO₃P)n
Smältpunkt: 600 °C
Densitet: 2,4 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134118
CAS nummer: 50813-16-6
EINECS: 256-779-4
Merck Index: 15, 8773
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Titration excess HCl 65 to 70 %
Chloride (Cl) ≤0.03 %
Sulfate (SO4) ≤0.1 %
Iron (Fe) ≤0.01 %