Kiselgel 2-5 mm, non toxic för torkning, with moisture indicator (orange gel)
Leverantör: Thermo Fisher Scientific
392030010 392030050 392032500
ACRO392030010EA 1625 SEK
ACRO392030010 ACRO392030050 ACRO392032500
Kiselgel 2-5 mm, non toxic för torkning, with moisture indicator (orange gel)
Kiselgel
Formel: SiO₂
Storage Temperature: Omgivande
MDL Number: MFCD00077645
CAS nummer: 7631-86-9
Order Now
Specifikation testresultat
Appearance Orange Beads
Bead size 2 to 5 mm
Color Orange (dry) to Colorless (wet)
Residual water ≤2 % (150°C)
Water adsorption ≥20 % (50% rH, 1 bar, 25°C)