trinatriumortofosfat, vattenfri, ren
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: tri-Sodium phosphate, Sodium phosphate tribasic, Trisodium phosphate
389810010 389810025 389810100
ACRO389810010EA 1775 SEK
ACRO389810010 ACRO389810025 ACRO389810100
trinatriumortofosfat, vattenfri, ren
trinatriumortofosfat
Formel: Na₃O₄P
MW: 163.94 g/mol
Smältpunkt: 73.3…76.7 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003510
CAS nummer: 7601-54-9
Merck Index: 12,08808
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Titration with HCl ≥96.0 %