Zinc(II) acetylacetonate (∼25% Zn)
Leverantör: Thermo Fisher Scientific
Varning
388770050 388771000 388775000
ACRO388770050EA 470 SEK
ACRO388770050 ACRO388771000 ACRO388775000
Zinc(II) acetylacetonate (∼25% Zn)
Zinc(II) acetylacetonate
Formel: C₁₀H₁₄O₄Zn
Kokpunkt: 129…131 °C (10 mmHg)
Smältpunkt: 124…126 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000035
CAS nummer: 14024-63-6
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Complexometric 23.5 to 26.0 % (Zinc)