Nickeldiklorid 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Nickel dichloride
387320500 387322500
ACRO387320500EA 1200 SEK
ACRO387320500 ACRO387322500
Nickeldiklorid 98%
Nickeldiklorid
Formel: Cl₂Ni
MW: 129,6 g/mol
Smältpunkt: 1001 °C
Densitet: 3,55 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011142
CAS nummer: 7718-54-9
UN: 3288
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Light orange to brown Powder
Titration Argentometric ≥97.5 %