Biphenyl-4,4'-dicarboxylic acid 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 4,4'-Biphenyldicarboxylic acid, [1,1'-Biphenyl]-4,4'-dicarboxylic acid
387300050 387300250
ACRO387300050EA 1925 SEK
ACRO387300050 ACRO387300250
Biphenyl-4,4'-dicarboxylic acid 98%
Biphenyl-4,4'-dicarboxylic acid
Formel: C₁₄H₁₀O₄
MW: 242,23 g/mol
Smältpunkt: 300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002554
CAS nummer: 787-70-2
Order Now
Specifikation testresultat
Appearance White to light beige Powder
Infrared spectrum Conforms
HPLC ≥97.5 %