trans-β-Nitrostyrene 97+%
Leverantör: Thermo Scientific
Varning
Synonyms: (E)-(2-Nitrovinyl)benzene, trans-1-Nitro-2-phenylethylene
386530050 386530250 386531000
ACRO386530050EA 595 SEK
ACRO386530050 ACRO386530250 ACRO386531000
trans-β-Nitrostyrene 97+%
trans-β-Nitrostyrene
Formel: C₈H₇NO₂
MW: 149,15 g/mol
Kokpunkt: 250…260 °C (760 mmHg)
Smältpunkt: 56…58 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00007402
CAS nummer: 5153-67-3
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Yellow Crystals
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥97.0 %