1-Allylpiperazine 99%
Leverantör: Thermo Scientific
|
Fara
|
384730010
384730050
ACRO384730010EA
545
SEK
ACRO384730010
ACRO384730050
1-Allylpiperazine 99%
1-Allylpiperazine
Formel:
C₇H₁₄N₂ MW: 126,2 g/mol Kokpunkt: 181…182 °C (760 mmHg) Densitet: 1 g/cm³ Flampunkt: 51,7 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00167970 CAS nummer: 13961-36-9 UN: 2734 ADR: 8,II |
Specifikation testresultat
Appearance | Clear light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4830 to 1.4850 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...