1H-Pyrazole-4-boronic acid pinacol ester 97+%
Leverantör: Thermo Fisher Scientific
Varning
382940010 382940050 382940250
ACRO382940010EA 1625 SEK
ACRO382940010 ACRO382940050 ACRO382940250
1H-Pyrazole-4-boronic acid pinacol ester 97+%
Formel: C₉H₁₅BN₂O₂
Smältpunkt: 125…130 °C
Storage Temperature: Kylskåp
MDL Number: MFCD03453063
CAS nummer: 269410-08-4
Order Now
Specifikation testresultat
Appearance Off-white to tan Powder
Infrared spectrum Conforms
GC ≥97.0 %