Propargyl acetate 97%
Leverantör: Thermo Scientific
Fara
Synonyms: Propargylacetate
380820050 380820250
ACRO380820050EA 680 SEK
ACRO380820050 ACRO380820250
Propargyl acetate 97%
Propargyl acetate
Formel: C₅H₆O₂
MW: 98,10 g/mol
Kokpunkt: 27…28 °C (8 mmHg)
Densitet: 0,989 g/cm³
Flampunkt: 33 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041601
CAS nummer: 627-09-8
UN: 1992
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4165 to 1.4195 (20°C, 589 nm)
GC ≥96.0 %