Lithium (trimethylsilyl)acetylide 0.5 M i THF, AcroSeal®
Leverantör: Thermo Scientific
Fara
Synonyms: lithium ethynyl(trimethyl)silane, LITHIUM(1+) ION (TRIMETHYLSILYL)ETHYNE, lithio(trimethylsilyl)acetylene
380651000 380658000
380651000.EA 3000 SEK
380651000. 380658000.
Lithium (trimethylsilyl)acetylide 0.5 M i THF, AcroSeal®
Lithium (trimethylsilyl)acetylide
Formel: C₅H₉LiSi
MW: 104,152 g/mol
Densitet: 0,88 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00075059
CAS nummer: 54655-07-1
UN: 2924
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear yellow to orange Solution
Titration with HClO4 0.40 to 0.60 M