4-Ethynylanisole 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (4-Methoxyphenyl)ethyne, (4-Methoxyphenyl)acetylene, 1-Ethynyl-4-methoxybenzene, 1-Methoxy-4-ethynylbenzene
379920010 379920050
ACRO379920010EA 1200 SEK
ACRO379920010 ACRO379920050
4-Ethynylanisole 97%
4-Ethynylanisole
Formel: C₉H₈O
Kokpunkt: 87…91 °C (11 mmHg)
Smältpunkt: 28…30 °C
Densitet: 1,019 g/cm³
Flampunkt: 82 °C
Storage Temperature: Omgivande
MDL Number: MFCD00168815
CAS nummer: 768-60-5
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to brown Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.5625 to 1.5675 (20°C, 589 nm)
GC ≥96.0 %