2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite 95%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Bis(diisopropylamino)(2-cyanoethoxy)phosphine, 3-((Bis(diisopropylamino)phosphino)oxy)propanenitrile
379740010 379740050 379740250
ACRO379740010EA 1925 SEK
ACRO379740010 ACRO379740050 ACRO379740250
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite 95%
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite
Formel: C₁₅H₃₂N₃OP
MW: 301.41 g/mol
Kokpunkt: 98…100 °C (5 mmHg)
Densitet: 0.949 g/cm³
Flampunkt: 60 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00012213
CAS nummer: 102691-36-1
UN: 1993
ADR: 3 III
Order Now
Specifikation testresultat
Appearance Clear to hazy colorless to yellow Liquid
Infrared spectrum Conforms
Titration with HClO4 ≥94 %