Benzo[b]furan-2-boronic acid 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Benzofuran-2-boronic acid, 2-Benzofurylboronic acid
378730010 378730050 378730250
ACRO378730010EA 765 SEK
ACRO378730010 ACRO378730050 ACRO378730250
Benzo[b]furan-2-boronic acid 97%
Benzo[b]furan-2-boronic acid
Formel: C₈H₇BO₃
Smältpunkt: 112…114 °C
Storage Temperature: Omgivande
MDL Number: MFCD00236019
CAS nummer: 98437-24-2
Order Now
Specifikation testresultat
Appearance White to yellow Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %