2-(Benzyloxy)acetaldehyde 95% stabilised
Leverantör: Thermo Scientific
Varning
Synonyms: (Benzyloxy)acetaldehyde, Benzyloxyacetaldehyde
376070010 376070050
ACRO376070010EA 1050 SEK
ACRO376070010 ACRO376070050
2-(Benzyloxy)acetaldehyde 95% stabilised
2-(Benzyloxy)acetaldehyde
Formel: C₉H₁₀O₂
MW: 150,177 g/mol
Kokpunkt: 118…120 °C (13 mmHg)
Densitet: 1,069 g/cm³
Flampunkt: 113 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00191779
CAS nummer: 60656-87-3
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
Stabilizer 0.5 % hydroquinone