cis-Epoxysuccinic acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: cis-2,3-Oxiranedicarboxylic Acid
372640050 372640250
ACRO372640050EA 2025 SEK
ACRO372640050 ACRO372640250
cis-Epoxysuccinic acid 97%
cis-Epoxysuccinic acid
Formel: C₄H₄O₅
MW: 132,07 g/mol
Smältpunkt: 133…135 °C
Storage Temperature: Omgivande
MDL Number: MFCD00191645
CAS nummer: 16533-72-5
Order Now
Specifikation testresultat
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH 96.0 to 104.0 %