Sodium cyanotrihydroborate 1M i THF, AcroSeal®
Leverantör: Thermo Scientific
Fara
Synonyms: Sodium cyanoborohydride
372451000 372458000
372451000.EA 1350 SEK
372451000. 372458000.
Sodium cyanotrihydroborate 1M i THF, AcroSeal®
Sodium cyanotrihydroborate
Formel: CH₃BNNa
MW: 62,84 g/mol
Smältpunkt: 241…243 °C
Densitet: 0,915 g/cm³
Flampunkt: –18 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003516
CAS nummer: 25895-60-7
UN: 3286
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear to slightly hazy colorless Solution (May develop some turbidity or precipitate)
Titration Iodimetric 0.95 to 1.10 M