Ammoniak 0.5 M 1,4-dioxane, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
368381000 368380010
368381000.EA 2050 SEK
368381000. 368380010.
Ammoniak 0.5 M 1,4-dioxane, AcroSeal®
Ammoniak
Formel: NH₃
MW: 17.03 g/mol
Kokpunkt: -33
Smältpunkt: -77
Densitet: 1.023 g/cm³
Flampunkt: 11 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00011418
CAS nummer: 7664-41-7
UN: 1993
ADR: 3 II
Merck Index: 13,00492
Order Now
Specifikation testresultat
Appearance Clear colorless Solution
Water ≤1 % (K.F.)
Strength 0.4 to 0.6 M